Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSN7b Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB291983 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291983 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291983 Supplier Novus Biologicals Supplier No. NB100461PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CSN7b Polyclonal specifically detects CSN7b in Human samples. It is validated for Western Blot.

Specifications

Antigen CSN7b
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias Arabidopsis, homolog) subunit 7B, COP9 constitutive photomorphogenic homolog subunit 7B (Arabidopsis), COP9 signalosome complex subunit 7b, CSN7BMGC111077, FLJ12612, Signalosome subunit 7b
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residues 225-264 (C-terminus) of human constitutive photomorphogenic homolog subunit 7B using the TrEMBL entry Q9H9Q2 (GeneID 64708).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Neurotransmission, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 64708
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.