Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSNK1A1L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179514
Description
CSNK1A1L Polyclonal specifically detects CSNK1A1L in Human samples. It is validated for Western Blot.Specifications
| CSNK1A1L | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| casein kinase 1, alpha 1-like, casein kinase I alpha S-like, casein kinase I isoform alpha-like, CK1, CKI-alpha-like, EC 2.7.11, EC 2.7.11.1, MGC33182 | |
| Rabbit | |
| 39 kDa | |
| 100 μL | |
| Signal Transduction | |
| 122011 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_660204 | |
| CSNK1A1L | |
| Synthetic peptide directed towards the middle region of human CSNK1A1LThe immunogen for this antibody is CSNK1A1L. Peptide sequence TLNHQYDYTFDWTMLKQKAAQQAASSSGQGQQAQTQTGKQTEKNKNNVKD. | |
| Affinity purified | |
| RUO | |
| Primary | |
| This product recognizes Human. Zebrafish: 79%; Dog: 77%; Pig: 77%; Rat: 77%; Horse: 77%; Mouse: 77%; Guinea pig: 77%. | |
| Human, Mouse, Rat, Canine, Equine, Guinea Pig, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction