Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSNK1A1L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | CSNK1A1L |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CSNK1A1L Polyclonal specifically detects CSNK1A1L in Human samples. It is validated for Western Blot.Specifications
| CSNK1A1L | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| casein kinase 1, alpha 1-like, casein kinase I alpha S-like, casein kinase I isoform alpha-like, CK1, CKI-alpha-like, EC 2.7.11, EC 2.7.11.1, MGC33182 | |
| CSNK1A1L | |
| IgG | |
| 39 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_660204 | |
| 122011 | |
| Synthetic peptide directed towards the middle region of human CSNK1A1LThe immunogen for this antibody is CSNK1A1L. Peptide sequence TLNHQYDYTFDWTMLKQKAAQQAASSSGQGQQAQTQTGKQTEKNKNNVKD. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title