Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSNK1A1L Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094439 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094439 20 μL
NB094438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094439 Supplier Novus Biologicals Supplier No. NBP30468720UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CSNK1A1L Polyclonal specifically detects CSNK1A1L in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CSNK1A1L
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias casein kinase 1, alpha 1-like, casein kinase I alpha S-like, casein kinase I isoform alpha-like, CK1, CKI-alpha-like, EC 2.7.11, EC 2.7.11.1, MGC33182
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-337 of human CSNK1A1L (NP_660204.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 122011
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.