Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSNK1A1L Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | CSNK1A1L |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
CSNK1A1L Polyclonal specifically detects CSNK1A1L in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CSNK1A1L | |
Polyclonal | |
Purified | |
RUO | |
Human | |
casein kinase 1, alpha 1-like, casein kinase I alpha S-like, casein kinase I isoform alpha-like, CK1, CKI-alpha-like, EC 2.7.11, EC 2.7.11.1, MGC33182 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-337 of human CSNK1A1L (NP_660204.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS with 50% glycerol, pH7.3. | |
122011 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title