Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSRP2BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596659 Supplier Invitrogen™ Supplier No. PA596659
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGIYGAKEGG ISRLPAGQAT YRTTCQDFRI LDRYQTSLPS RKGFRHQTTK FLYRLVGSED MAVDQSIVSP YTSRILKPYI RRDYETKPPK LQLLSQIRSH LHRSDPHWTP EPDAPLDYCY VRPNHIPTIN SMCQEFFWPG IDLSECLQYP DFSVVVLYKK VIIAFGFMVP DVKYNEAYIS FLFVHPEWRR AGIATFMIYH LIQTCMGKDV TLHVSASNPA MLLYQKFGFK TEEYVLDFYD KYYPLESTEC KHAFFLRLRR; Positive Samples: HL-60, A375, SW480, Mouse kidney, Mouse heart, Mouse brain; Cellular Location: Cytoplasm, Nucleus.

CSRP2 is a protein containing two LIM domains, which are double zinc finger motifs found in proteins of diverse function. CSRP2 and some related proteins are thought to act as protein adapters, bridging two or more proteins to form a larger protein complex. The protein encoded by this gene binds to one of the LIM domains of CSRP2 and contains an acetyltransferase domain. Although the encoded protein has been detected in the cytoplasm, it is predominantly a nuclear protein. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSRP2BP
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KAT14
Gene Accession No. Q9H8E8
Gene Alias 2510008M08Rik; ADA2A-containing complex subunit 2; ATAC component 2 homolog; ATAC2; AU023459; CRP2 binding partner; CRP2 binding protein; CRP2-binding partner; CRP2BP; CSRP2 binding protein; CSRP2-binding protein; Csrp2bp; cysteine and glycine-rich protein 2 binding protein; cysteine-rich protein 2 binding protein; cysteine-rich protein 2-binding protein; D2Ertd473e; D2Wsu131e; dJ717M23.1; E430020F17; Kat14; Lysine acetyltransferase 14; PRO1194
Gene Symbols KAT14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 523-782 of human CSRP2BP (NP_0653973).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57325
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.