Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSRP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7088208 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6539 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6539 Supplier Proteintech Group Inc Supplier No. 107211AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cysteine and glycine-rich protein 3 (CSRP3) is a member of the CSRP family of genes encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in CRP3 is found in a group of proteins with critical functions in gene regulation, cell growth, and somatic differentiation. Very strongly similar to rat MLP; contains zinc-finger LIM domains. Other genes in the family include CSRP1 and CSRP2.

Specifications

Antigen CSRP3
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CSRP3
Gene Accession No. P50461, P50462, P50463
Gene Alias Cardiac LIM protein, CLP, CMD1M, CMH12, CRP3, CSRP3, Cysteine rich protein 3, LIM domain protein, cardiac, LMO4, MLP, Muscle LIM protein
Gene Symbols CSRP3
Host Species Rabbit
Immunogen CSRP3 Fusion Protein Ag1136
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117505, 13009, 8048
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.