Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSTF2T Antibody (PCRP-CSTF2T-1A3), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB390492 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390492 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390492 Supplier Novus Biologicals Supplier No. NBP308807AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CSTF2T Monoclonal antibody specifically detects CSTF2T in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen CSTF2T
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-CSTF2T-1A3
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CF-1 64 kDa subunit tau, CF-1 64 kDa subunit tau variant, cleavage stimulation factor 64 kDa subunit tau, Cleavage stimulation factor 64 kDa subunit tau variant, cleavage stimulation factor subunit 2 tau, cleavage stimulation factor subunit 2 tau variant, cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau variant, CSTF 64 kDa subunit tau variant, CstF-64T, DKFZp434C1013, KIAA0689CSTF 64 kDa subunit tau, tauCstF-64
Host Species Mouse
Immunogen Recombinant full-length human CSTF2T protein (Uniprot: Q9H0L4)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23283
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.