Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSTF50 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501559890
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
Catalog No. Quantity
50-155-9890 10 μL
1 options

Catalog No. 50-155-9890

Supplier: Bethyl Laboratories, Inc A301251AT

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Orangutan

This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5′ UTR, but encoding the same protein, have been found for this gene.

Specifications

Antigen CSTF50
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CSTF1
Gene Accession No. Q05048
Gene Alias CstF-50, CstFp50
Gene Symbols CSTF1
Host Species Rabbit
Immunogen Between 381 and 431
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1477
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.