Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTAG1B Antibody (OTI2B6), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB278489 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB278489 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB278489 Supplier Novus Biologicals Supplier No. NBP270485MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CTAG1B Monoclonal antibody specifically detects CTAG1B in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CTAG1B
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2B6
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Autoimmunogenic cancer/testis antigen NY-ESO-1, cancer antigen 3, cancer/testis antigen 1, cancer/testis antigen 1B, Cancer/testis antigen 6.1, CT6.1L antigen family member 2, CTAG1CTAG1A, ESO1CTAG, LAGE2, LAGE2A, LAGE2BLAGE-2, New York esophageal squamous cell carcinoma 1, NY-ESO-1
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human CTAG1B expression vector
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1485
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.