Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CtBP1 Antibody (4D6), Biotin, Novus Biologicals™
SDP

Catalog No. NBP174040B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP174040B 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP174040B Supplier Novus Biologicals Supplier No. NBP174040B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CtBP1 Monoclonal specifically detects CtBP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CtBP1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 4D6
Conjugate Biotin
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias BARS, brefeldin A-ribosylated substrate, CTBP, CtBP1, C-terminal binding protein 1, C-terminal-binding protein 1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.158, MGC104684
Gene Symbols CTBP1
Host Species Mouse
Immunogen Recombinant human CTBP1 (1-440aa) purified from E. coli
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1487
Test Specificity Anti-human CTBP1 mAb, clone 4D6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CTBP1 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.