Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTBP1-DT Antibody (OTI2C6), HRP, Novus Biologicals™
SDP

Catalog No. NB288233 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288233 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288233 Supplier Novus Biologicals Supplier No. NBP272333H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CTBP1-DT Monoclonal antibody specifically detects CTBP1-DT in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF

Specifications

Antigen CTBP1-DT
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone OTI2C6
Conjugate HRP
Dilution Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF-ready
Formulation PBS
Gene Alias CTBP1 antisense RNA 1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-186 of human C4orf42(NP_443093) produced in E.coil.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92070
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.