Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTBP1-DT Antibody (OTI4F2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP272334A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272334A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272334A30 Supplier Novus Biologicals Supplier No. NBP272334AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CTBP1-DT Monoclonal specifically detects CTBP1-DT in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen CTBP1-DT
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI4F2
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CTBP1 antisense RNA 1
Gene Symbols CTBP1-DT
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-186 of human C4orf42 (NP_443093) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 92070
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.