Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTBP1-DT Antibody (OTI4F2), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP272334R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272334R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272334R Supplier Novus Biologicals Supplier No. NBP272334R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CTBP1-DT Monoclonal specifically detects CTBP1-DT in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen CTBP1-DT
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI4F2
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CTBP1 antisense RNA 1
Gene Symbols CTBP1-DT
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-186 of human C4orf42 (NP_443093) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 92070.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.