Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CtBP1 Rabbit anti-Human, Mouse, Rat, Clone: 1I7G6, Novus Biologicals™
SDP

Catalog No. NB165921 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165921 20 μg
NB165920 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB165921 Supplier Novus Biologicals Supplier No. NBP31541220UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CtBP1 Monoclonal antibody specifically detects CtBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen CtBP1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 1I7G6
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BARS, brefeldin A-ribosylated substrate, CTBP, CtBP1, C-terminal binding protein 1, C-terminal-binding protein 1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.158, MGC104684
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 301-440 of human CtBP1 (Q13363).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1487
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.