Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CtBP1 Rabbit anti-Human, Mouse, Rat, Clone: JG39-73, Novus Biologicals™
SDP

Catalog No. NBP275453 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275453 Supplier Novus Biologicals Supplier No. NBP275453

Rabbit Monoclonal Antibody

CtBP1 Monoclonal specifically detects CtBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen CtBP1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JG39-73
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias BARS, brefeldin A-ribosylated substrate, CTBP, CtBP1, C-terminal binding protein 1, C-terminal-binding protein 1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.158, MGC104684
Gene Symbols CTBP1
Host Species Rabbit
Immunogen Recombinant protein within Human CtBP1 aa 261-440/ 440. (SwissProt: Q13363 Human; SwissProt: Q9Z2F5 Rat)
Molecular Weight of Antigen 47 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cell Cycle and Replication, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1487.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.