Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTBP2 Antibody (PCRP-CTBP2-2D11), Alexa Fluor™ 405 - C-terminus, Novus Biologicals™
SDP

Catalog No. NB386056 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386056 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386056 Supplier Novus Biologicals Supplier No. NBP308550AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CTBP2 Monoclonal antibody specifically detects CTBP2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen CTBP2
Applications Flow Cytometry, ELISA, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-CTBP2-2D11
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias CtBP2, C-terminal binding protein 2, 10ribeye, C-terminal-binding protein 2
Host Species Mouse
Immunogen Recombinant full-length human CTBP2 protein (Uniprot: P56545)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1488
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.