Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTCF Recombinant Rabbit Monoclonal Antibody, AbFlex™, Active Motif™

Rabbit Recombinant Monoclonal Antibody
$108.45 - $482.00
Specifications
Antigen | CTCF |
---|---|
Concentration | 1 mg/mL |
Applications | ChIP sequencing (ChIP-seq), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3800
![]() ![]() |
Active Motif
91286 |
10 μg |
Each for $108.45
|
|
|||||
50-199-3799
![]() ![]() |
Active Motif
91285 |
100 μg |
Each for $482.00
|
|
|||||
Description
Transcriptional factor CTCF is a ubiquitously expressed 11-zinc finger domain containing, DNA-binding nuclear phosphoprotein that is involved in multiple aspects of normal gene regulation including transcriptional repression and activation, gene silencing, chromatin insulation and regulation of imprinted sites. CTCF uses different combinations of ZF domains and interacts with the CCCTC motif on the DNA. It can bind to HAT- and HDAC-containing complexes thereby influencing transcriptional activation or repression respectively. It is a multifunctional protein involved in the transactivation of APPB gene, silencing of c-myc gene, and insulation of human beta-globin and DMI myotonic dystrophy locus and imprinting control of IGF2 and H19 regions. CTCF has also been shown to regulate the transcription of Pax6 and IRAK2 promoter thereby opening a therapeutic avenue. Mutations in this gene have been associated with invasive breast cancers, prostate cancers and Wilms' tumors.Specifications
CTCF | |
ChIP sequencing (ChIP-seq), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P49711 | |
10664 | |
This antibody was raised against a peptide within human CTCF. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
CTCF |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
11 zinc finger transcriptional repressor; 11-zinc finger protein; AW108038; BOS_17095; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein); CTCF; ctcf protein; CTCFL paralog; im:7152666; MRD21; OTTHUMP00000197610; Transcriptional repressor CTCF; Unknown (protein for IMAGE:8125283) | |
CTCF | |
IgG | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title