Learn More
Description
Specifications
Specifications
| Antigen | CTDSPL |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | O15194 |
| Gene Alias | C3orf8, Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 3, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase-like, CTD small phosphatase-like protein, CTDSP-like, EC 3.1.3, HYA22, NIF1, NIFL, NIF-like protein, NLI-interacting factor 1, Nuclear LIM interactor-interacting factor 1, Protein YA22, PSR1, RB protein serine phosphatase from chromosome 3, RBSP3EC 3.1.3.16, SCP3chromosome 3 open reading frame 8, Small CTD phosphatase 3, Small C-terminal domain phosphatase 3, YA22 |
| Gene Symbols | CTDSPL |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
