Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTDSPL Antibody, Novus Biologicals™
SDP

Catalog No. NBP153169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP153169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP153169 Supplier Novus Biologicals Supplier No. NBP153169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CTDSPL Polyclonal specifically detects CTDSPL in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CTDSPL
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q3ZTU0
Gene Alias C3orf8, Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 3, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase-like, CTD small phosphatase-like protein, CTDSP-like, EC 3.1.3, HYA22, NIF1, NIFL, NIF-like protein, NLI-interacting factor 1, Nuclear LIM interactor-interacting factor 1, Protein YA22, PSR1, RB protein serine phosphatase from chromosome 3, RBSP3EC 3.1.3.16, SCP3chromosome 3 open reading frame 8, Small CTD phosphatase 3, Small C-terminal domain phosphatase 3, YA22
Gene Symbols CTDSPL
Host Species Rabbit
Immunogen Synthetic peptides corresponding to CTDSPL(CTD (carboxy-terminal domain) small phosphatase-like) The peptide sequence was selected from the N terminal of CTDSPL. Peptide sequence CCFRDYNVEAPPPSSPSVLPPLVEENGGLQKPPAKYLLPEVTVLDYGKKC.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10217
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.