Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTGF/CCN2 Antibody (18E7), Biotin, Novus Biologicals™
SDP

Catalog No. NB162808 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162808 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162808 Supplier Novus Biologicals Supplier No. NBP260235B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CTGF/CCN2 Monoclonal antibody specifically detects CTGF/CCN2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen CTGF/CCN2
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 18E7.
Conjugate Biotin
Formulation PBS
Gene Alias CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2
Host Species Mouse
Immunogen Recombinant human CTGF/CCN2 (27-349 aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cytokine Research, Growth and Development, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 1490
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.