Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTGF/CCN2 Antibody (CL5339), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$382.00 - $646.00

Specifications

Antigen CTGF/CCN2
Clone CL5339
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP261415
SDP
View Documents
Novus Biologicals
NBP261415
100 μL
Each for $646.00
Only null left
Add to Cart
 
NB393113
SDP
View Documents
Novus Biologicals
NBP26141525UL
25 μL
Each for $382.00
Only null left
Add to Cart
 
Description

Description

CTGF/CCN2 Monoclonal specifically detects CTGF/CCN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CTGF/CCN2
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Unconjugated
Mouse
Human, Mouse, Rat
1490
This antibody was developed against a recombinant protein corresponding to amino acids: LPSPDCPFPRRVKLPGKCCEEWVCDEPKDQTVVGPALAAYRLEDTFGPDPTMIRANCLVQTTEWSACSKTCGMG
Primary
Specificity of human CTGF/CCN2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
CL5339
Monoclonal
Purified
Angiogenesis, Cancer, Cell Biology, Cytokine Research, Growth and Development, Immunology, Innate Immunity
CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2
CCN2
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.