Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTGF/CCN2 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB100724UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100724UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100724UV Supplier Novus Biologicals Supplier No. NB100724UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CTGF/CCN2 Polyclonal specifically detects CTGF/CCN2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen CTGF/CCN2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2
Gene Symbols CCN2
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of human CTGF (between residues 299-349). [UniProtNo. P29279]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1490
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.