Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTGF/CCN2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP266426 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266426 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266426 Supplier Novus Biologicals Supplier No. NBP266426
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CTGF/CCN2 Polyclonal specifically detects CTGF/CCN2 in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.

Specifications

Antigen CTGF/CCN2
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 5ug/ml, Peptide ELISA detection limit 1:10000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2
Gene Symbols CCN2
Host Species Goat
Immunogen Peptide with sequence C-SLYYRKMYGDMA, from the C Terminus of the protein sequence according to NP_001892.
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Angiogenesis, Cancer, Cell Biology, Cytokine Research, Growth and Development, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 1490
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.