Learn More
CTGF Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$250.00 - $319.00
Specifications
Antigen | CTGF |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P25250UG
![]() |
Invitrogen™
500P25250UG |
50 μg |
Each for $250.00
|
![]() |
|||||
500P252100
![]() |
Invitrogen™
500P252100UG |
100 μg |
Each for $319.00
|
|
|||||
Description
AA Sequence of recombinant protein: GKKCIRTPKI SKPIKFELSG CTSMKTYRAK FCGVCTDGRC CTPHRTTTLP VEFKCPDGEV MKKNMMFIKT CACHYNCPGD NDIFESLYYR KMYGDMA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human CTGF. Anti-Human CTGF-specific antibody was purified by affinity chromatography employing an immobilized Human CTGF matrix. Sandwich ELISA: To detect Human CTGF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human CTGF (500-P252Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human CTGF. Western Blot: To detect Human CTGF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human CTGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Connective Tissue Growth Factor (CTGF), also called CCN2, is a member of the CCN family. CTGF is a secreted protein produced by umbilical veins and vascular endothelial cells. CTGF possesses an Insulin-like Growth Factor (IGF)-binding domain, a thrombospondin type 1 domain, and a cysteine knot region. CTGF plays important roles in the proliferation and differentiation of chondrocytes, induces angiogenesis, and promotes cell adhesion in fibroblasts, endothelial, and epithelial cells.Specifications
CTGF | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P29279 | |
1490 | |
E.coli-derived Recombinant Human CTGF. | |
Antigen affinity chromatography | |
CCN2 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
2-Nov; betaIG-M2; CCN family member 2; Ccn2; Cellular communication network factor 2; connective tissue growth factor; connective tissue growth factor precursor; Connective tissue growth factor precursor (CTGF) (FISP-12 protein) (Hypertrophic chondrocyte-specific protein 24); connective tissue growth-related protein; CTGF; CTGF protein; CTGRP; fibroblast inducible secreated protein; fibroblast inducible secreted protein; Fisp12; fisp-12; Hcs24; H-CTGF; hypertrophic chondrocyte-specific gene product 24; hypertrophic chondrocyte-specific protein 24; IBP-8; IGF-binding protein 8; IGFBP8; IGFBP-8; Insulin-like growth factor-binding protein 8; MGC102839; NOV2; Protein FISP-12; similar to connective tissue growth factor precursor | |
CCN2 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.