Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTHRC1 Antibody (166), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB110290 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB110290 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB110290 Supplier Novus Biologicals Supplier No. NBP289947AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CTHRC1 Monoclonal specifically detects CTHRC1 in Human samples. It is validated for Western Blot.

Specifications

Antigen CTHRC1
Applications Western Blot
Classification Monoclonal
Clone 166
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CTHRC1 (NP_612464.1; Met1-Lys243).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115908
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.