Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ctip1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB60258A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60258A594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB60258A594 Supplier Novus Biologicals Supplier No. NB600258AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ctip1 Polyclonal specifically detects Ctip1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Ctip1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias B-cell CLL/lymphoma 11A, B-cell CLL/lymphoma 11A (zinc finger protein), B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2, B-cell lymphoma/leukemia 11A, BCL-11A, BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1, BCL11A-S, BCL11A-XL, C2H2-type zinc finger protein, COUP-TF-interacting protein 1, CTIP1ecotropic viral integration site 9, ecotropic viral integration site 9 homolog, Ecotropic viral integration site 9 protein homolog, EVI-9, EVI9FLJ34997, HBFQTL5, KIAA1809BCL11A-L, Zinc finger protein 856, ZNF856FLJ10173
Gene Symbols BCL11A
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human B-cell CLL/Lymphoma 11A using the numbering given in Swiss-Prot entry Q9H165 (GeneID 53335).
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 53335
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.