Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ctip1 Antibody (BLR073G), Novus Biologicals™
SDP

Catalog No. NB000890 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB000890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000890 Supplier Novus Biologicals Supplier No. NBP278710
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Ctip1 Monoclonal specifically detects Ctip1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.

Specifications

Antigen Ctip1
Applications Western Blot, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone BLR073G
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry, Immunoprecipitation 6 uL/1 mg lysate
Formulation Borate Buffered Saline (BBS) pH 8.2. with 0.09% Sodium Azide
Gene Alias B-cell CLL/lymphoma 11A, B-cell CLL/lymphoma 11A (zinc finger protein), B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2, B-cell lymphoma/leukemia 11A, BCL-11A, BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1, BCL11A-S, BCL11A-XL, C2H2-type zinc finger protein, COUP-TF-interacting protein 1, CTIP1ecotropic viral integration site 9, ecotropic viral integration site 9 homolog, Ecotropic viral integration site 9 protein homolog, EVI-9, EVI9FLJ34997, HBFQTL5, KIAA1809BCL11A-L, Zinc finger protein 856, ZNF856FLJ10173
Gene Symbols BCL11A
Host Species Rabbit
Immunogen A peptide representing a region between residue 1 and 50.
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μL
Research Discipline Apoptosis, Cancer, Inhibitors Activators and Regulators, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 53335
Target Species Human, Mouse
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.