Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTLA-4 Antibody (1B8) - BSA Free, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | CTLA-4 |
---|---|
Clone | 1B8 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:100, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 |
Applications | Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Flow Cytometry, Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018515
![]() |
Novus Biologicals
NBP1410000.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NBP14100001
![]() |
Novus Biologicals
NBP1410000.1MG |
0.1 mg |
Each for $499.50
|
|
|||||
Description
CTLA-4 Monoclonal specifically detects CTLA-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
CTLA-4 | |
1.0 mg/mL | |
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Flow Cytometry, Western Blot, ELISA | |
Unconjugated | |
Armenian Hamster | |
Signal Transduction | |
PBS (pH 7.4) with 0.09% Sodium Azide | |
CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
CTLA4 | |
IgG | |
Protein G purified | |
This antibody recognises mouse CD152, commonly known as CTLA-4 (cytotoxic T-lymphocyte protein 4), a structural homolog for CD28. CD152 binds the same ligands as CD28, namely CD80 (B7.1) and CD86 (B7.2), but with much higher affinity. However, while binding of CD28 enhances T cell function, binding of CD152 inhibits it. CD152 is not detected on naive T-cells and is induced upon T-cell activation. |
1B8 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:100, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
P09793 | |
1493 | |
Extracellular portion of mouse CD152 fused to a mouse IgG2a. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title