Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTLA-4 Antibody (2188B) [DyLight 488], Novus Biologicals™
SDP

Catalog No. NB372724 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372724 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372724 Supplier Novus Biologicals Supplier No. FAB3253K
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CTLA-4 Monoclonal specifically detects CTLA-4 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen CTLA-4
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 2188B
Conjugate DyLight 488
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CTLA-4, Ala37-Phe162, Accession # P16410
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1493
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.