Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTLA-4 Antibody (37B3D8.2F2), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB337092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337092 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337092 Supplier Novus Biologicals Supplier No. NBP231143MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen CTLA-4
Applications Western Blot
Classification Monoclonal
Clone 37B3D8.2F2
Conjugate mFluor Violet 610 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Host Species Mouse
Immunogen Mouse CTLA4
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1493
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.