Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTLA-4 Antibody (9H10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB151850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151850 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151850 Supplier Novus Biologicals Supplier No. NBP311965AF350
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen CTLA-4
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 9H10
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Host Species Armenian Hamster
Immunogen This antibody was raised by immunising Syrian hamsters with Staphylococcus A bacteria coated in CTLA-4, isolating B cells from the immunised hamsters and fusing these with the P3X3.Ag8.653 myeloma line to produce stable hybridomas.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1493
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.