Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTLA-4 Antibody (WKH203), PE
SDP

Catalog No. NB10063254Q Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063254Q 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063254Q Supplier Novus Biologicals Supplier No. NB10063254PE

Mouse Monoclonal Antibody

CTLA-4 Monoclonal specifically detects CTLA-4 in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen CTLA-4
Applications Flow Cytometry
Classification Monoclonal
Clone WKH203
Conjugate PE
Dilution Flow Cytometry
Gene Alias CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Gene Symbols CTLA4
Host Species Mouse
Immunogen Purified rCTLA-4hIg fusion protein.
Purification Method Protein G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1493
Test Specificity NB100-63254 recognizes rat CD152, also known as cytotoxic-T-lymphocyte antigen-4 (CTLA-4), which is similar in structure to CD28 and also binds ligands CD80 and CD86. CD152 is expressed by activated T-lymphocytes. Studies suggest that CD152 is also expressed by regulatory T-lymphocytes.
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.