Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTNNA3 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB057899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057899 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057899 Supplier Novus Biologicals Supplier No. NBP299083G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CTNNA3 Polyclonal specifically detects CTNNA3 in Human samples. It is validated for Western Blot.

Specifications

Antigen CTNNA3
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Alpha T-catenin, alpha-catenin-like protein, alpha-T-catenin, Cadherin-associated protein, catenin (cadherin-associated protein), alpha 3, catenin alpha-3, MGC26194, MGC75041, VR22
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human CTNNA3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29119
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.