Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTP synthase Antibody (13B6), Novus Biologicals™
SDP

Catalog No. NB297696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297696 50 μL
NB297697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297696 Supplier Novus Biologicals Supplier No. NBP32664050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CTP synthase Monoclonal antibody specifically detects CTP synthase in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen CTP synthase
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 13B6
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias CTP synthase, CTP synthase 1, CTP synthetase 1, cytidine 5-prime triphosphate synthetase, cytidine 5'-triphosphate synthetase, EC 6.3.4.2, UTP--ammonia ligase 1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human CTP synthase [UniProt P17812]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1503
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.