Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTR2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP10199A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP10199A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP10199A30 Supplier Novus Biologicals Supplier No. NBP105199AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CTR2 Polyclonal specifically detects CTR2 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen CTR2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Copper transporter 2, COPT2SLC13A2, CTR2Solute carrier family 31 member 2, hCTR2probable low affinity copper uptake protein 2, solute carrier family 31 (copper transporters), member 2
Gene Symbols SLC31A2
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human CTR2 (within residues 50-100). [Swiss-ProtNo. O15432]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1318
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.