Learn More
Description
Specifications
Specifications
| Antigen | CTRP7 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | K14-S |
| Concentration | 3.6mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:5000, ELISA 1:20000-1:50000 |
| Formulation | 20mM Tris/HCl (pH 8.0) and 10 mg/ml BSA with 0.05% Sodium Azide |
| Gene Accession No. | Q9BXJ2 |
| Gene Alias | C1q and tumor necrosis factor related protein 7, complement-c1q tumor necrosis factor-related protein 7, CTRP7complement C1q tumor necrosis factor-related protein 7, ZACRP7 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
