Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTRP7 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB357720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357720 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357720 Supplier Novus Biologicals Supplier No. NBP176635MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CTRP7 Polyclonal antibody specifically detects CTRP7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen CTRP7
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C1q and tumor necrosis factor related protein 7, complement-c1q tumor necrosis factor-related protein 7, CTRP7complement C1q tumor necrosis factor-related protein 7, ZACRP7
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human CTRP7. The immunogen is located within the first 50 amino acids of CTRP7. Amino Acid Squence: QPRGNQLKGENYSPR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 114905
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.