Learn More
Invitrogen™ CTU1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total ATPBD3.
Specifications
Specifications
| Antigen | CTU1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CTU1 |
| Gene Accession No. | Q7Z7A3 |
| Gene Alias | ATP binding domain 3; Atpbd3; ATP-binding domain-containing protein 3; BC005752; Cancer-associated gene protein; cancer-associated protein; CTU1; Cytoplasmic tRNA 2-thiolation protein 1; cytoplasmic tRNA adenylyltransferase 1; cytosolic thiouridylase subunit 1; cytosolic thiouridylase subunit 1 homolog; cytosolic thiouridylase subunit 1 homolog (S. pombe); NCS6 |
| Gene Symbols | CTU1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.