Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CU CPT 22
GSA_VAGREENER_CHOICE

Catalog No. p-4817472
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective TLR1/2 inhibitor

Selective toll-like receptor 1/2 (TLR1/2) inhibitor (IC50 = 0.58 μM). Exhibits no activity at TLR2/TLR6, TLR3, TLR4 and TLR7 or a panel of 10 representative kinases. Inhibits the release of proinflammatory cytokines TNF-α and IL-1β.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1416324-85-0
Molecular Formula C19H22O7

Specifications

Quantity 10 mg
Formula Weight 362.37
Purity >98%
Molecular Weight (g/mol) 362.378
Chemical Name or Material CU CPT 22
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.