Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CU CPT 4a
GREENER_CHOICE

Catalog No. 488310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-831-0 10 mg
48-835-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-831-0 Supplier Tocris Bioscience™ Supplier No. 4883/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective TLR3 inhibitor

Selective TLR3 inhibitor (IC50 = 3.44 μM in RAW 264.7 cells); suppresses downstream signaling pathways mediated by the TLR3/dsRNA complex, inhibiting TNF-α and IL-1β production in whole cells. Reduces death of crypt cells and improves gastrointestinal syndrome in mice.

Chemical Identifiers

CAS 1279713-77-7
Molecular Formula C18H13ClFNO3S

Specifications

Quantity 10 mg
Formula Weight 377.82
Molecular Weight (g/mol) 377.814
Chemical Name or Material CU CPT 4a
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.