Learn More
Sartorius Cubis™ II 116S High-Capacity Advanced Micro Balance
GREENER_CHOICE
Catalog No. 01675480 Shop All Sartorius Products
Click to view available options
Model:
HWL Configuration with Draft Shield
ION Configuration with Draft Shield
MDS Configuration with Draft Shield
QP99 Configuration with Draft Shield
QP99 ION Configuration with Draft Shield
QP99 MDS Configuration with Draft Shield
Standard Configuration
Standard Configuration with Draft Shield
The Cubis™ II range of high-capacity micro balances with a maximum load between 32 g and 111 g and a readability between 0.001 mg and 0.002 mg provide the ideal choice for a broad range of applications.
  • Ultra-high resolution with up to 61 million weighing steps (digits)
  • End-to-end data integrity. 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management
  • Cleanability, part of compliance
  • Error-free operation. Individual QApp workflows, motorized auto-leveling
  • Allow upgrades for hardware features like automated draft shields or built-in ionizer
  • Optional inner draft shield for best weighing performance
  • Housing: Stainless steel 1.4401, 1.4404, Aluminum, Plastic PBT, PA, Float glass Optiwhite, Control Unit: Aluminum, painted, Plastic PBT, PP, Float glass
TRUSTED_SUSTAINABILITY
Specifications
Connection Type USB A, USB B, USB C
Capacity 111 g
Display Type Color TFT Touchscreen
Readability 0.002 mg
Material Stainless Steel
Model Standard Configuration
Stabilization Time 3.5 sec.
Type Micro Balance
Electrical Requirements 100/240 V
Legal for Trade No
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.