Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Cubis II™ Essential High Capacity Balance
GSA_VAGREENER_CHOICE

Catalog No. p-7229308 Shop All Sartorius Products
Click to view available options
Model:
36200S
36201P
Readability:
0.1 g, 1 g
1 g
Capacity:
36.2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cubis II™ Essential High Capacity Balance offers a high level of weighing accuracy with a large, high-contrast touch display that features factory-installed essential weighing applications for easy operation.

  • Top Features of the Cubis II™ Essential High-Capacity Balance
  • Stainless steel weigh pan
  • Convenient essential user interface
  • 1 g readability
  • Popular Applications for the Cubis II™ Essential High-Capacity Balance: Large sample weighing, Chemical analysis and consumer care

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Connection Type USB A USB B USB C RS-232 9 Pin peripheral connector for Sartorius accessories
Capacity 36.2 kg
Display Type Touchscreen
Readability 0.1 g, 1 g
Material Lead (CAS 7439-92-1): This substance is present as an ingredient in brass alloys. It complies with the limit of 0.4% by weight of EU Directive 2011/65 / EU (RoHS) in these alloys. Bisphenol A (CAS 80-05-7): Bisphenol A is the raw material for the plastic polymers polycarbonate and polysulfone. Sartorius uses these two polymers in some plastic components. Bisphenol A is firmly bound in the structure of the plastic. If used as intented, no adverse health effects are to be expected.
Model 36201P
Stabilization Time 1.5 sec.
Type High Capacity Toploader
Electrical Requirements CR2030
Legal for Trade Class III
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.