Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUEDC2 Antibody (003), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB110011 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB110011 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB110011 Supplier Novus Biologicals Supplier No. NBP289902AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CUEDC2 Monoclonal specifically detects CUEDC2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CUEDC2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CUEDC2.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79004
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.