Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUEDC2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224639 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224639 0.1 mg
NBP224639SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224639 Supplier Novus Biologicals Supplier No. NBP224639
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CUEDC2 Polyclonal specifically detects CUEDC2 in Bovine, Canine, Equine, Human, Mouse, Pig, Primate samples. It is validated for Western Blot.

Specifications

Antigen CUEDC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1.0 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9H467
Gene Alias bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491
Gene Symbols CUEDC2
Host Species Rabbit
Immunogen A portion of amino acid 250-287 of human CUEDC2 was used as the immunogen.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79004
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rat and mouse, 83% homologous in tetradon, pig, xenopus and pufferfish, and 77% homologous in chicken.
Target Species Bovine, Canine, Equine, Human, Mouse, Pig, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.