Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUEDC2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | CUEDC2 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224639SS
![]() |
Novus Biologicals
NBP224639SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224639
![]() |
Novus Biologicals
NBP224639 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
CUEDC2 Polyclonal specifically detects CUEDC2 in Human, Mouse, Porcine, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot.Specifications
CUEDC2 | |
Western Blot 0.5-1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491 | |
CUEDC2 | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in human, rat and mouse, 83% homologous in tetradon, pig, xenopus and pufferfish, and 77% homologous in chicken. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Pig, Bovine, Canine, Equine, Primate | |
Q9H467 | |
79004 | |
A portion of amino acid 250-287 of human CUEDC2 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title