Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUGBP1/CELF1 Antibody (3B1), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB20316A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB20316A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB20316A350 Supplier Novus Biologicals Supplier No. NB200316AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CUGBP1/CELF1 Monoclonal specifically detects CUGBP1/CELF1 in Human, Mouse, Rat, Porcine, Bovine, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CUGBP1/CELF1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3B1
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BRUNOL250 kDa nuclear polyadenylated RNA-binding protein, bruno-like 2, Bruno-like protein 2, CUG RNA-binding protein, CUG triplet repeat, RNA binding protein 1, CUG triplet repeat, RNA-binding protein 1, CUG-BP, CUG-BP- and ETR-3-like factor 1, CUGBP Elav-like family member 1, CUGBP, Elav-like family member 1, CUGBP1CUG triplet repeat RNA-binding protein 1, CUGBPCELF-1, Deadenylation factor CUG-BP, EDEN-BP, EDEN-BP homolog, embryo deadenylation element binding protein, Embryo deadenylation element-binding protein homolog, hNab50, NAB50CUG-BP1, NAPOR, nuclear polyadenylated RNA-binding protein, 50-kD, RNA-binding protein BRUNOL-2
Gene Symbols CELF1
Host Species Mouse
Immunogen Full-length human CUGBP1 [UniProtNo. Q92879]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10658
Target Species Human, Mouse, Rat, Pig, Bovine, Primate, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.