Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUGBP2 Antibody (PCRP-CELF2-1E4), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB157580 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB157580 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB157580 Supplier Novus Biologicals Supplier No. NBP314271AF594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CUGBP2 Monoclonal antibody specifically detects CUGBP2 in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen CUGBP2
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-CELF2-1E4
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias BRUNOL3ELAV-type RNA-binding protein 3, Bruno-like protein 3, CELF-2, CUG triplet repeat RNA-binding protein 2, CUG triplet repeat, RNA binding protein 2, CUG triplet repeat, RNA-binding protein 2, CUG-BP- and ETR-3-like factor 2, CUGBP Elav-like family member 2, CUGBP, Elav-like family member 2, CUG-BP2, CUGBP2hNAPOR, Etr-3, ETR3RNA-binding protein BRUNOL-3, NAPOR, NAPOR-2, Neuroblastoma apoptosis-related RNA-binding protein
Host Species Mouse
Immunogen Recombinant full-length human CUGBP2 protein (Uniprot: O95319)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10659
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.