Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUGBP2 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 129211AP150UL

Description
CUG-BP1 and CUG-BP2 are proteins that bind specifically to (CUG)8 oligonucleotides in vitro. While CUG-BP1 has the major binding activity in normal cells, nuclear CUG-BP2 binding activity increases in DM cells. Both CUG-BP1 and CUG-BP2 are isoforms of a novel heterogeneous nuclear ribonucleoprotein (hnRNP), hNab50. CUG-BP1, an RNA CUG triplet repeat binding protein, regulates splicing and translation of various RNAs. Expansion of RNA CUG repeats in the DMPK in DM is associated with alterations in binding activity of CUG-BP1 as well as alterations in the translation of the C/EBPb transcription factor. CUG-BP1 is an important regulator of initiation from different AUG codons of C/EBPb mRNA.Specifications
CUGBP2 | |
Polyclonal | |
Unconjugated | |
CELF2 | |
Bruno like protein 3, BRUNOL3, CELF 2, CELF2, CUG BP2, CUGBP2, ETR 3, ETR3, hNAPOR, NAPOR, RNA binding protein BRUNOL 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10659, 14007, 29428, 373117 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95319, Q6P0B1, Q792H5, Q9Z0H4 | |
celf2 | |
CUGBP2 Fusion Protein Ag3594 | |
150 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction