Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUL4B Antibody (OTI1C4), PE, Novus Biologicals™
SDP

Catalog No. NB288925 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288925 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288925 Supplier Novus Biologicals Supplier No. NBP271373PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CUL4B Monoclonal antibody specifically detects CUL4B in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen CUL4B
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1C4
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias CUL-4B, cullin 4B, cullin-4B, DKFZp686F1470, KIAA0695, MRXHF2, MRXS15, MRXSC, SFM2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-300 of human CUL4B (NP_003579) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8450
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.